Order Entry
Export
ContactUsLinkComponent
 
Anti-CD16/CD32 Rat Monoclonal Antibody [clone: 2.4G2] (FITC)
 
undefined
Anti-CD16/CD32 Rat Monoclonal Antibody [clone: 2.4G2] (FITC)

 

  • Catalog No:
  • STMC60161FI.1
  • STMC60161FI
  • Antigen name:
  • CD16/CD32
  • CD16/CD32
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Clone:
  • 2.4G2
  • 2.4G2
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 14130/14131
  • 14130/14131
  • Isotype:
  • IgG2b kappa
  • IgG2b kappa