Order Entry
Export
ContactUsLinkComponent
Anti-CD11b Mouse Monoclonal Antibody [clone: ICRF44] (FITC)
Anti-CD11b Mouse Monoclonal Antibody [clone: ICRF44] (FITC)
Certificates
undefined
Anti-CD11b Mouse Monoclonal Antibody [clone: ICRF44] (FITC)

Specifications

  • Catalog No:
  • STMC60040FI
  • STMC60040FI.1
  • Antigen name:
  • CD11b
  • CD11b
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Clone:
  • ICRF44
  • ICRF44
  • Reactivity:
  • Baboon, Pig, Cynomolgus monkey, Common marmoset, Rhesus monkey, Chimpanzee, Human
  • Baboon, Pig, Cynomolgus monkey, Common marmoset, Rhesus monkey, Chimpanzee, Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 3684
  • 3684
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa

Specifications