Specifications
- Antigen name:Epithelial Cell
- Clonality:Monoclonal
- Clone:5E11.3.1
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Gene ID:4072
- Size:100 µg
- Pk:0,1 mL
Specifications
About this item
Mouse monoclonal IgG1 antibody against human epithelial cells, unconjugated.
- Mammary cells
The 5E11.3.1 antibody reacts with an unidentified marker expressed on the surface of most luminal cells of normal human mammary epithelium. Weak labeling (or no labeling) is observed within the basal cell compartment of the mammary epithelium. The 5E11.3.1 antibody gives variable labeling on malignant breast epithelium. Positive labeling is observed on the following cell lines: MCF7, CAMA, T47D, BT-10, BT-20, COLO 205, and WiDr. Labeling is negative on human bone marrow cells, blood cells, lymphoid and mesenchymal tissues, mammary and marrow fibroblast cultures, and human embryonic stem cell line H9.
Type: Anti-Epithelial Cell Mouse Monoclonal Antibody [clone: 5E11.3.1]
Antigen: Epithelial Cell
Clonality: Monoclonal
Clone: 5E11.3.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human