- Antigen name:TRA-2-49
- Clonality:Monoclonal
- Clone:TRA-2-49/6E
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Pig,Rabbit,Non-human primates,Human,Cat
- Gene ID:249
- Size:100 μg
- Pk:0,2 mL
Mouse monoclonal IgG1 antibody against human, chimpanzee, gibbon TRA-2-49 (tissue non-specific alkaline phosphatase), unconjugated.
- Pluripotent stem cells
The TRA-2-49/6E antibody reacts with the tissue non-specific isoform of alkaline phosphatase (AP), which is expressed at high levels in undifferentiated pluripotent stem cells such as human and mouse embryonic stem (ES) cells, as well as induced pluripotent stem (iPS) and embryonal carcinoma (EC) cells. This isozyme of AP is the same as that expressed by liver, bone and kidney cells. Levels of AP decrease upon differentiation and, thus, the TRA-2-49/6E antibody can be used to monitor the differentiation status of pluripotent stem cells. The TRA-2-49/6E antibody does not cross-react with other isozymes of AP, which differ through post-translational modification, and reportedly does not recognise AP from cow, dog, goat, Guinea pig, hamster, mouse, rat or sheep, or Old World Monkeys such as the rhesus monkey, baboon and African green monkey.
Type:
Antigen: TRA-2-49
Clonality: Monoclonal
Clone: TRA-2-49/6E
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Cat, Human, Non-Human Primate, Pig, Rabbit