Order Entry
Export
ContactUsLinkComponent
Anti-CD8a Rat Monoclonal Antibody [clone: 53-6.7] (Biotinilated)
Anti-CD8a Rat Monoclonal Antibody [clone: 53-6.7] (Biotinilated)
Certificates
undefined
Anti-CD8a Rat Monoclonal Antibody [clone: 53-6.7] (Biotinilated)

Specifications

  • Catalog No:
  • STMC60023BT.1
  • STMC60023BT
  • Antigen name:
  • CD8a
  • CD8a
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Biotin
  • Biotin
  • Clone:
  • 53-6.7
  • 53-6.7
  • Reactivity:
  • Frog, Mouse
  • Frog, Mouse
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 12525
  • 12525
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa

Specifications