Order Entry
Export
ContactUsLinkComponent
Anti-CD19 Rat Monoclonal Antibody [clone: 6D5] (Alexa Fluor® 488)
Anti-CD19 Rat Monoclonal Antibody [clone: 6D5] (Alexa Fluor® 488)
Certificates
undefined
Anti-CD19 Rat Monoclonal Antibody [clone: 6D5] (Alexa Fluor® 488)

Specifications

  • Catalog No:
  • STMC60006AD
  • STMC60006AD.1
  • Antigen name:
  • CD19
  • CD19
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Alexa Fluor® 488
  • Alexa Fluor® 488
  • Clone:
  • 6D5
  • 6D5
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 12478
  • 12478
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa

Specifications