Order Entry
Export
ContactUsLinkComponent
Human Recombinant LIF
Human Recombinant LIF
Certificates
undefined
Human Recombinant LIF

Specifications

  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Protein/peptide name:
    LIF
  • Purity:
    >95%

Specifications