Order Entry
Export
Orders LinkContactUsLinkComponent
Human Recombinant IL-10

Human Recombinant IL-10

Human Recombinant IL-10
Certificates
Human Recombinant IL-10

3 Options Available Below

Product Details & Documents

Interleukin 10 (IL-10) is the founding member of the IL-10 family of class II cytokines. All of the IL-10 cytokine family members have a four-helix bundle consisting of α-helical folds. Upon binding to its receptor, IL-10 activates signaling through JAK1 and STAT3. It is produced by dendritic cells, macrophages, and CD4+ T cells, as well as mast cells, NK cells, neutrophils, and B cells, under specific stimulating conditions (Saraiva and O'Garra). IL-10 can inhibit the expression of pro-inflammatory cytokines and promote healing processes, and is important for the function of regulatory T cells. IL-10 also enhances B cell proliferation, immunoglobulin secretion, and class II MHC expression, while IL-10 produced by macrophages inhibits activation of neighboring macrophages, thus allowing a level of self-regulation (Ouyang et al.).

 : Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • STMC78024.1
  • STMC78024
  • STMC78024.2
  • Protein/peptide name:
  • IL-10
  • IL-10
  • IL-10
  • Protein/peptide type:
  • Recombinant
  • Recombinant
  • Recombinant
  • Species:
  • Human
  • Human
  • Human
  • Purity:
  • ≥95%
  • ≥95%
  • ≥95%

Specifications

Product Family Options

Product Information

  • Pk
    Availability
    Price
  • 10 µG
    Log in for availability
    Log in for availability
    1 * 10 µG
    1 * 10 µG - 275,00 €
    275,00 €
    1 * 10 µG
       275,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    STMC78024.1
    Protein/peptide name
    IL-10
    Protein/peptide type
    Recombinant
    Species
    Human
    Purity
    ≥95%
  • 50 µG
    Log in for availability
    Log in for availability
    1 * 50 µG
    1 * 50 µG - 832,00 €
    832,00 €
    1 * 50 µG
       832,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    STMC78024
    Protein/peptide name
    IL-10
    Protein/peptide type
    Recombinant
    Species
    Human
    Purity
    ≥95%
  • 1.000 µG
    Log in for availability
    Log in for availability
    1 * 1.000 µG
    1 * 1.000 µG - 7 220,00 €
    7 220,00 €
    1 * 1.000 µG
       7 220,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    STMC78024.2
    Protein/peptide name
    IL-10
    Protein/peptide type
    Recombinant
    Species
    Human
    Purity
    ≥95%