Human Recombinant EGF
Supplier: STEMCELL Technologies
Certificates
About this item
Epidermal growth factor (EGF) is characterized by high affinity binding to various EGF receptors (EGFRs) and the production of mitogenic responses (Carpenter and Cohen). EGF promotes EGFR dimerization, resulting in activation of downstream pathways including PI3K, ERK1/2, JAK/STAT, β-catenin, and calcium signaling. EGF is secreted by the gut-associated salivary and Brunner’s glands, is found in a variety of body fluids, and stimulates cell proliferation and differentiation in rodent and neonatal human intestine (Wright et al.). Central nervous system stem cells also proliferate in response to the EGF stimulus (Reynolds and Weiss).
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Specifications
- Protein/peptide type:Recombinant
- Species:Human
- Protein/peptide name:EGF
- Purity:>95%