Order Entry
Export
ContactUsLinkComponent
Mouse Recombinant EGF
Mouse Recombinant EGF
Certificates
undefined
Mouse Recombinant EGF

Specifications

  • Protein/peptide type:
    Recombinant
  • Species:
    Mouse
  • Protein/peptide name:
    EGF
  • Purity:
    ≥95 %

Specifications