Human Recombinant FGF-9
Catalog # STMC78161.1
Supplier: STEMCELL Technologies
CAS Number:
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Species:Human
- Protein/peptide name:FGF-9
- Purity:≥95%
Specifications
About this item
Fibroblast growth factor (FGF) 9 is a pleiotropic member of the FGF family. FGFs are heparin-dependent regulators of cell proliferation and differentiation (Itoh and Ornitz). FGF-9 signals by binding FGFR3 (IIIb) and the IIIc splice variants of the FGFR1, FGFR2, and FGFR3 receptors (Santos-Ocampo et al.; Mohammadi et al.; Plotnikov et al.). In mice, deletion of FGF-9 is lethal and expression of FGF-9 affects development of the skeleton, cerebellum, lungs, heart, vascular system, digestive tract, and testes (Colvin et al.; Harada et al.; Lin et al.). Altered levels of FGF-9 have also been reported in various types of human cancers (Leushacke et al.; Abdel-Rahman et al.).
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').