Order Entry
Export
ContactUsLinkComponent
Human Recombinant IL-12
Human Recombinant IL-12
Catalog # STMC78027.2
CAS Number:  
undefined
Human Recombinant IL-12
Catalog # STMC78027.2
CAS Number:  

Specifications

  • Pk:
    1.000 µG
  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Protein/peptide name:
    IL-12
  • Purity:
    ≥95%

Specifications

About this item

Interleukin 12 (IL-12p70) is a heterodimeric cytokine composed of p35 and p40 subunits. IL-12 is produced by monocytes, macrophages, dendritic cells, neutrophils, and B cells in response to bacterial products and cytokines such as IFN-γ. The IL-12 receptor is expressed on T, NK, and dendritic cells. Upon binding, IL-12 initiates signaling via the JAK/STAT signaling pathway and stimulates NK, B, and T cells to produce IFN-γ (Watford et al.). It also regulates cytokine synthesis, proliferation of T and NK cells, and stimulates differentiation of CD4+ and CD8+ T cells (Germann and Rüde). Mice that are deficient in IL-12 are susceptible to many intracellular pathogens and have impaired IFN-γ secretion, Th1 differentiation, and NK cytolytic activity; however, Th2 development and IL-4 production are enhanced (Watford et al.).

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').