Order Entry
Export
Orders LinkContactUsLinkComponent
Human (Actin) Peptide Pool

Human (Actin) Peptide Pool

1 AU
Catalog # STMC100-1411
Human (Actin) Peptide Pool
Catalog # STMC100-1411
CAS Number:  
Human (Actin) Peptide Pool
Human (Actin) Peptide Pool

1 AU

CAS Number:  
Price
556,00 €
Log in for availability
1 * 1 AU
Product Details & Documents

Human (Actin) peptide pool is a lyophilised mixture of 92 peptides from human actin, alpha skeletal muscle, and consists of 15-mer peptides with 11-amino-acid overlaps that cover amino acids 1 to 377 on actin. It is frequently used as a negative control. Actin, alpha skeletal muscle, is part of the highly conserved actin protein family and is the predominant isoform found in adult skeletal muscle (Laing et al.). Peptide pools are useful for a broad range of applications, including vaccine development, immunological research, and diagnostic assay development.

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Specifications

Specifications

  • Pk:
    1 AU
  • Cat. No.:
    STMC100-1411
  • Protein/peptide name:
    (Actin) Peptide Pool
  • Purity:
    Average 70%
  • Source:
    Human cells

Specifications