Order Entry
Export
ContactUsLinkComponent
Human Recombinant IL-7
Human Recombinant IL-7
Certificates
undefined
Human Recombinant IL-7

Specifications

  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Protein synonyms:
    Interleukin-7|LP-1|Lymphopoietin 1|Pre-B cell factor
  • Protein/peptide name:
    IL-7
  • Purity:
    ≥95%

Specifications