Order Entry
Export
ContactUsLinkComponent
Human Recombinant TNF-alpha
Human Recombinant TNF-alpha
Certificates
undefined
Human Recombinant TNF-alpha

Specifications

  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Protein/peptide name:
    TNF-alpha
  • Purity:
    ≥95%

Specifications