Order Entry
Export
Orders LinkContactUsLinkComponent
Human Recombinant FGF-18

Human Recombinant FGF-18

Human Recombinant FGF-18
Certificates
Human Recombinant FGF-18

2 Options Available Below

Product Details & Documents

Fibroblast growth factor 18 (FGF-18) is a growth factor and member of the FGF subfamily. FGF-18 is most similar to FGF-8 and FGF-17, and is secreted in adult lung and developing tissues (Ohbayashi et al.). FGF-18 signals through FGF receptor 3 (FGFR3) to regulate proliferation, differentiation, and matrix production of articular and growth plate chondrocytes in vivo and in vitro (Davidson et al.). FGF-18 has skeletal functions and protects articular cartilage by gene expression profiling and regulates proliferation and differentiation of midline cerebellar structures (Mori et al.). Also, recombinant human FGF-18 has been shown to effectively regulate hair growth (Song et al.).

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • STMC78041
  • STMC78041.1
  • Protein/peptide name:
  • FGF-18
  • FGF-18
  • Protein/peptide type:
  • Recombinant
  • Recombinant
  • Species:
  • Human
  • Human
  • Purity:
  • ≥95%
  • ≥95%

Specifications

Product Family Options

Product Information

  • Pk
    Availability
    Price
  • 10 µG
    Log in for availability
    Log in for availability
    1 * 10 µG
    1 * 10 µG - 147,00 €
    147,00 €
    1 * 10 µG
       147,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    STMC78041
    Protein/peptide name
    FGF-18
    Protein/peptide type
    Recombinant
    Species
    Human
    Purity
    ≥95%
  • 50 µG
    Log in for availability
    Log in for availability
    1 * 50 µG
    1 * 50 µG

    Specifications:
    More
    Specifications
    Cat. No.
    STMC78041.1
    Protein/peptide name
    FGF-18
    Protein/peptide type
    Recombinant
    Species
    Human
    Purity
    ≥95%