Order Entry
Export
ContactUsLinkComponent
Human Recombinant 15-PGDH, His tag
Human Recombinant 15-PGDH, His tag
Catalog # STMC100-1329
CAS Number:  
undefined
Human Recombinant 15-PGDH, His tag
Catalog # STMC100-1329
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Protein synonyms:
    PGDH|PGDH1|PHOAR1|SDR36C1
  • Protein/peptide name:
    15-PGDH
  • Purity:
    ≥89%

Specifications

About this item

Modulate prostaglandin metabolism with 15-hydroxyprostaglandin dehydrogenase (15-PGDH). 15-PGDH catalyses the reversible oxidation of the 15-hydroxyl group in prostaglandins, resulting in inactivated metabolites (Ensor and Tai), it can act on a variety of prostaglandins as substrates in a NAD+ dependent manner (Cho et al.). As prostaglandins can have a range of effects on cellular processes, 15-PGDH is of considerable importance in drug development. In vitro and in vivo studies suggest 15-PGDH has tumor suppressive activity and is downregulated in certain cancers (Na et al.).15-PGDH belongs to the short-chain dehydrogenases/reductases (SDR) family; its primary structure indicates 20% homology with other SDRs, with some conserved amino acid residues (Krook et al.). This protein contains a His-residue tag at the carboxyl end of the polypeptide chain. For consistency and reproducibility across your applications, apo M from STEMCELL comes lyophilised with ≥89% purity.

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').