Human Recombinant Midkine
Product Details & Documents
Midkine is a member of a unique family of heparin-binding growth factors that are structurally different from other fibroblast growth factors (Muramatsu; Takada et al.). Midkine is a proinflammatory cytokine, promoting the migration of leukocytes, fibrinolysis, and acting as a chemotactic agent towards neutrophils (Muramatsu; Said et al.; Takada et al.). It also regulates growth, differentiation, and development during the midgestion stage of embryogenesis, and promotes angiogenesis (Muramatsu; Said et al.; Takada et al.). The protein structure consists of three antiparallel β-sheets and is highly conserved between species (Muramatsu; Takada et al.). While the exact signal pathway is not known, proposed pathways include promoting LRP, inhibiting Src kinase, activating paxillin and STAT1α, activating PI2 and MAP kinases, suppressing caspases, binding to α6β1-integrin and tetraspanin, activating FAK, phosphorylating STAT3, suppressing STAT5 phosphorylation, activating ALK, activating PI3 kinase and transcription of NFkB, binding to neuroglycan C or nucleolin, and binding to eIF3 (Muramatsu). In cultured cells, midkine influences growth and survival of neural precursor cells, synthesis of cytokines from endothelial and renal epithelial cells, and promotes synthesis of extracellular matrices from fibroblasts (Muramatsu; Takada et al.).
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Specifications
Specifications
- Pk:20 µG
- Cat. No.:STMC100-0933
- Protein/peptide type:Recombinant
- Protein/peptide name:Midkine
- Protein synonyms:Amphiregulin-associated protein|ARAP|MDK|MEK|Midgestation and kidney protein|MK1|MKARAP|NEGF2|NEGF2FLJ27379|Neurite outgrowth-promoting factor 2|Neurite outgrowth-promoting protein
- Purity:≥92%
- Species:Human



