Anti-BLOS3 Rabbit Polyclonal Antibody
Catalog # NOVUNBP303343-100
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen symbol:BLOS3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Format:BSA-free
- Antigen synonyms:biogenesis of lysosome-related organelles complex 1 subunit 3|biogenesis of lysosomal organelles complex-1, subunit 3|HPS8|RP
- Purity:Affinity purified
- Storage buffer:PBS with 50% glycerol, pH 7.3
- Storage temperature:Store at –20 °C. Avoid freeze-thaw cycles.
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1 - 202 of human BLOS3 (NP_997715.1)
- Pk:100 µl
Specifications
About this item
Immunocytochemistry/ Immunofluorescence 1:50 - 1:200; Immunohistochemistry 1:50 - 1:200; Immunohistochemistry-Paraffin.
Type: Primary
Antigen: BLOS3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat