Order Entry
Export
ContactUsLinkComponent
3,6-Dichloro-1,2-benzenedithiol 95%, Sigma-Aldrich®
Corrosive
Toxic
Health
Environment
3,6-Dichloro-1,2-benzenedithiol 95%, Sigma-Aldrich®
Catalog # 442828-1G
CAS Number:  
Corrosive
Toxic
Health
Environment
3,6-Dichloro-1,2-benzenedithiol 95%, Sigma-Aldrich®
Catalog # 442828-1G
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance (Colour):
    Off White to Green to Beige and Faint Orange to Orange
  • Appearance (Form):
    Powder or Crystal or Chunk(s)
  • Infrared spectrum:
    Conforms to Structure
  • Proton NMR spectrum:
    Conforms to Structure
  • Titration by AgNO3, After Oxygen Combustion:
    94.0 - 106.0%

Specification Test Results

About this item

 :
 :211.13 g/mol
UN:2811
 :GHS05
 :GHS06
 :GHS08
 :GHS09
Melting Pt:58...60 °C (lit.)
Storage Temperature:Room Temperature
ADR:6.1,II
CAS Number:87314-49-6
Formula:Cl₂C₆H₂(SH)₂
MDL Number:MFCD00216665