Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
Microscope Objectives
Lenses for Stemi 2000
Lenses for Stemi 2000
:
ZEISS
Lenses for Stemi 2000
:
ZEISS
Lenses for Stemi 2000
Product Family Options
Description
Magnification
Working distance
630-0360
Front lens
0,3×
285 mm
1 * 1 items
-
+
1 * 1 items
-
+
630-0361
Front lens
0,40×
210 mm
1 * 1 items
-
+
1 * 1 items
-
+
630-2398
Front lens
1,25×
60 mm
1 * 1 items
-
+
1 * 1 items
-
+