Order Entry
Export
ContactUsLinkComponent
 
Toxic
3,3′-Diethylthiacyanine iodide ~97% (dye content), Sigma-Aldrich®
Catalog # 390410-1G
CAS Number:  
undefined
Toxic
3,3′-Diethylthiacyanine iodide ~97% (dye content), Sigma-Aldrich®
Catalog # 390410-1G
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance (Colour), Yellow to Dark Green:
    Conforms to Requirements
  • Appearance (Form), Powder or Crystals:
    Conforms to Requirement
  • Infrared spectrum:
    Conforms to Structure
  • Wavelength, 0.004 g/L, Ethanol:
    420 - 428 nm
  • Extinction Coefficient:
    ≥65000
  • Carbon:
    47.2 - 50.6%
  • Nitrogen:
    5.7 - 6.2%
  • Purity (HPLC):
    ≥96.5%

Specification Test Results

About this item

 :
EINECS:218-593-1
UN:2811
 :GHS06
Melting Pt:>300 °C (lit.)
Storage Temperature:Room Temperature
ADR:6.1,III
 :M-Clarity™ quality level = MQ100
 :466.40 g/mol
CAS Number:2197-01-5
Formula:C₁₉H₁₉IN₂S₂
MDL Number:MFCD00050309