Diisobutylaluminium hydride 1 mol/l in cyclohexane, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:1191-15-7
:
Formula:C₈H₁₉Al
MDL Number:MFCD00008928
:142.22 g/mol
ADR:4.3,I
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:214-729-9
UN:3399