
Eosin B, pure, certified
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:548-24-3
:
Formula:C₂₀H₆Br₂N₂Na₂O₉
MDL Number:MFCD00005041
:624.06 g/mol
Merck Index:12,03638
Melting Pt:>310 °C (dec.)
EINECS:208-943-1
:GHS05
Storage Temperature:Ambient
Specification Test Results
- Appearance:Dark brown to dark red-purple Crystalline powder
- Infrared spectrum:Conforms
- Dye content:≥85 %
- UV Lambda max:511 to 520 nm (in water + 1% Na2CO3)
- Absorption Ratio P-15/P+15:0.96 to 1.22
- BSC Certification:Passes