Order Entry
Export
Orders LinkContactUsLinkComponent
Eosin B

Eosin B

Eosin B
Certificates
Eosin B

2 Options Available Below

Product Details & Documents

CAS No.: 548-24-3 Molecular Formula: C20H6Br2N2Na2O9 Formula Weight: 624.08 MDL No.: MFCD00005041

Synonyms:  Eosin bluish, 2,7-Dinitro-4,5-dibromofluorescein, Eosin B disodium salt, Sodium 2-(4,5-dibromo-2,7-dinitro-6-oxido-3-oxo-3H-xanthen-9-yl)benzoate, 2-(4,5-dibromo-3,6-dihydroxy-2,7-dinitroxanthen-9-yl)-benzoic acid disodium salt, Acid Red 91
  • CAS Number:548-24-3
  • Formula:C₂₀H₆Br₂N₂Na₂O₉
  • MDL Number:MFCD00005041
  • Molecular Weight:624.06 g/mol
  • Merck Index:12,03638
  • Melting Pt:> 310 °C
  • Storage Temperature:Ambient
  • EINECS:208-943-1
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 45,50 €
    45,50 €
    1 * 25 g
       45,50 €

  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 124,00 €
    124,00 €
    1 * 100 g
       124,00 €