Thiamine hydrochloride
Supplier: Roth Carl
Certificates
About this item
CAS Number:67-03-8
:
Formula:C₁₂H₁₈Cl₂N₄OS
MDL Number:MFCD00012780
:337.27 g/mol
Merck Index:13,09366
Melting Pt:252 °C
Density:1.401 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-641-8