
Neocuproine hemihydrate 99+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:34302-69-7
:
Formula:C₂₈H₂₆N₄O
MDL Number:MFCD00149306
:434.53 g/mol
Merck Index:13,06477
Melting Pt:158…163 °C
EINECS:207-601-9
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to beige Powder
- Infrared spectrum:Conforms
- Melting point:159 to 164 °C
- Titration with HClO4:99.0 to 101.0 %