Reichstein's substance S ≥98%
R0500-1G
:
- Pack type:Poly bottle
- Pk:1 g
- Seq:3047
- Environmentally Preferable:
- Appearance (Colour):White to Off-White
- Appearance (Form):Powder
- Infrared Spectrum:Conforms to Structure
- Purity (HPLC):≥98 %
- Solubility (Colour):Colorless to Faint Yellow
- Solubility (Turbidity)
50 mg/mL, EtOH:Clear - Recommended Retest Period:6 Years
: 17alpha,21-Dihydroxy-4-pregnene-3,20-dione, Reichstein's Substance S, 11-Deoxy-17-hydroxycorticosterone
:
:346.47 g/mol
:208 °C (dec., lit.)
:M-Clarity™ quality level = MQ200
:Room Temperature
:152-58-9
:C₂₁H₃₀O₄
:MFCD00003662