Pepsin
Supplier: Spectrum Chemical
About this item
Commonly used in biochemistry.
2 Options Available Below
Product Details & Documents
Commonly used in biochemistry.
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- SPCMP1571-100GM
- SPCMP1571-500GM
- Enzyme name:
- Pepsin
- Pepsin
- CAS No.:
- 9001-75-6
- 9001-75-6
- Form:
- Powder
- Powder
- Storage conditions:
- Refrigerator
- Refrigerator
- Grade:
- EP
- EP
- Tested applications:
- for biochemistry
- for biochemistry
Specifications
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:Small Business Enterprise
More
Specifications
Cat. No.SPCMP1571-100GMEnzyme namePepsinCAS No.9001-75-6FormPowderStorage conditionsRefrigeratorGradeEPTested applicationsfor biochemistrySmall Business Enterprise - Specifications:
More
Specifications
Cat. No.SPCMP1571-500GMEnzyme namePepsinCAS No.9001-75-6FormPowderStorage conditionsRefrigeratorGradeEPTested applicationsfor biochemistry



