BIT 9500 Serum Substitute
Catalog # STMC09500
Supplier: STEMCELL Technologies
CAS Number:
Specifications
- Pk:100 mL
- Description:BIT 9500 serum substitute
Specifications
About this item
Supplement for serum-free cell culture media.
BIT 9500 Serum Substitute has been developed for use in applications where a serum-free culture medium of defined composition is required. This product contains bovine serum albumin (BSA), insulin, and transferrin (BIT) in Iscove’s MDM. It contains pre-screened batches of BSA that have been selected to support the optimal growth of human hematopoietic progenitor cells in serum-free media formulations. It is also suitable for the culture of mouse hematopoietic progenitor cells in serum-free conditions.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').