ContactUsLinkComponent
MethoCult™-SF H4236 Medium
MethoCult™-SF H4236 Medium
Catalog # STMC04236
CAS Number:  
undefined
MethoCult™-SF H4236 Medium
Catalog # STMC04236
CAS Number:  
Specifications
  • Pk:
    80 mL
  • Description:
    MethoCult™ SF H4236
Specifications
About this item
Serum-free methylcellulose-based medium without cytokines for human cells.
MethoCult™ SF H4236 is recommended as a base medium for the culture of human cells in defined serum-free conditions, and to detect and quantify hematopoietic progenitor cells in human bone marrow (BM), mobilised peripheral blood (MPB), peripheral blood (PB), and cord blood (CB) samples using CFU assays. This formulation allows for the addition of an exogenous source of erythropoietin (EPO) and other cytokines, and is ideal for testing cytokine effects where the presence of fetal bovine serum is not desired.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').