ContactUsLinkComponent
MethoCult™-GF M3534 Medium
  STMC03534
 :  
undefined
MethoCult™-GF M3534 Medium
  STMC03534
 :  
  • Pk:
    100 mL
  • Description:
    MethoCult™ GF M3534
Methylcellulose-based medium with recombinant cytokines (without EPO) for mouse myeloid progenitor cells.
MethoCult™ GF M3534 is optimised for the growth and enumeration of granulocyte-macrophage progenitor cells (CFU-GM, CFU-G, CFU-M) in colony-forming unit (CFU) assays of mouse bone marrow, spleen, peripheral blood, and fetal liver cells. MethoCult™ M3534 does not support the growth of erythroid progenitor cells (BFU-E and CFU-E) as it does not contain erythropoietin (EPO). This formulation is compatible with STEMvision™ software for automated colony counting of mouse bone marrow CFU assays.
 : Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').