Orders LinkContactUsLinkComponent
ClonaCell™-HY Medium B
ClonaCell™-HY Medium B
ClonaCell™-HY medium B, 500 mL
Catalog # STMC03802
ClonaCell™-HY Medium B
Catalog # STMC03802
CAS Number:  
ClonaCell™-HY Medium B
ClonaCell™-HY Medium B
ClonaCell™-HY medium B, 500 mL
CAS Number:  
Price
341,00 €
Log in for availability
1 * 500 mL
Product Details & Documents
Hybridoma fusion medium (serum-free).
ClonaCell™-HY medium B is a serum-free liquid medium used to wash lymphocytes and parental myeloma cells to remove serum prior to fusion and during the fusion process. This medium has been verified for use in mouse and rat hybridoma development and reportedly is compatible for production of hybridomas using lymphocytes from a variety of host animals including human, mouse, rat, and hamster.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Specifications
Specifications
  • Pk:
    500 mL
  • Description:
    ClonaCell™-HY medium B
Specifications