trans-4-Stilbenemethanol
1 g
Catalog # 262013-1G
Supplier: SIGMA ALDRICH MICROSCOPY
trans-4-Stilbenemethanol
Catalog # 262013-1G
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number: 101093-37-2
Jump to:
Product Details & Documents
Synonyms: 4-Hydroxymethylstilbene
- CAS Number:101093-37-2
- Formula:C₁₅H₁₄O
- MDL Number:MFCD00011548
- Molecular Weight:210.27 g/mol
- Melting Pt:155...160 °C (lit.)
- :M-Clarity™ quality level = MQ100
- Storage Temperature:Room Temperature
Specifications
Specification Test Results
- Appearance (Colour):White to Yellow
- Appearance (Form):Powder or Crystals
- Infrared spectrum:Conforms to Structure
- Purity (GC):≥96.5%
Specifications
- Pk:1 g



