ContactUsLinkComponent
Septum Stopper, Sleeve Type
Septum Stopper, Sleeve Type
Catalog # VWRU89029-091
Supplier:  VWR Collection
New Product
Septum Stopper, Sleeve Type
Catalog # VWRU89029-091
Supplier:  VWR Collection
New Product
Specifications
  • Colour:
    White
  • Description:
    Stopper, sleeve type, for ²⁹/₂₆ to ²⁹/₄₂ outer joints
  • Pk:
    12
Specifications
About this item
Sleeve type stopper with hollow plugs and sleeve extensions.
  • Available in red or white rubber
The sleeve extensions fold down over the neck of a vessel and tubing. The diaphragm can be punctured with a syringe needle, re-sealing after the needle is withdrawn.