Anti-CD14 Mouse Monoclonal Antibody [clone: 61D3] (iFluor® 430)
Supplier: AAT Bioquest
Certificates
About this item
61D3 is an anti-human monoclonal antibody that targets the CD14 antigen.
- Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Caution: For research use only.
Type: Primary
Antigen: CD14
Clonality: Monoclonal
Clone: 61D3
Conjugation: iFluor® 430
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Specifications
- Catalog No:
- AATB10141030
- AATB10141031
- Antigen symbol:
- CD14
- CD14
- Antigen name:
- CD14 antigen
- CD14 antigen
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- iFluor® 430
- iFluor® 430
- Clone:
- 61D3
- 61D3
- Reactivity:
- Human
- Host:
- Mouse
- Mouse
- ImmunoFluorescence:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Purification:
- Affinity chromatography
- Affinity chromatography
- Cross adsorption:
- No
- No
- Storage temperature:
- Refrigerated (2...8 °C), minimise light exposure
- Refrigerated (2...8 °C), minimise light exposure
- Shipping temperature:
- 20
- 20