Anti-CD8a Mouse Monoclonal Antibody [clone: SK1]
Catalog # AATB10081000
Supplier: AAT BIOQUEST
Specifications
- Antibody type:Primary
- Antigen name:CD8a antigen
- Antigen symbol:CD8a
- Clonality:Monoclonal
- Clone:SK1
- Flow cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Reactivity:Human
- Cross adsorption:No
- Storage temperature:Refrigerated (2...8 °C); Minimise light exposure
- Shipping temperature:20
- Purification:Affinity chromatography
- Pack type:Vial
- Pk:100 µG
Specifications
About this item
SK1 is an anti-human monoclonal antibody that is specific for the CD8a antigen.
- Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Caution: For research use only.
Type: Primary
Antigen: CD8a antigen
Clonality: Monoclonal
Clone: SK1
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human