Order Entry
Export
ContactUsLinkComponent
Anti-CD44 Rat Monoclonal Antibody [clone: IM7] (iFluor® 660)
Anti-CD44 Rat Monoclonal Antibody [clone: IM7] (iFluor® 660)
Supplier:  AAT Bioquest
Certificates
undefined
Anti-CD44 Rat Monoclonal Antibody [clone: IM7] (iFluor® 660)
Supplier:  AAT Bioquest

Specifications

  • Catalog No:
  • AATB104410G1
  • AATB104410G0
  • Antigen symbol:
  • CD44
  • CD44
  • Antigen name:
  • CD44 antigen
  • CD44 antigen
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • iFluor® 660
  • iFluor® 660
  • Clone:
  • IM7
  • IM7
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rat
  • Rat
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Purification:
  • Affinity chromatography
  • Affinity chromatography
  • Cross adsorption:
  • No
  • No
  • Storage temperature:
  • 2...8 °C with minimised light exposure. Do not freeze.
  • 2...8 °C with minimised light exposure. Do not freeze.
  • Shipping temperature:
  • 20
  • 20

Specifications