Anti-CD4 Mouse Monoclonal Antibody [clone: OKT-4] (iFluor® 594)
Catalog # AATB100430C0
Supplier: AAT BIOQUEST
Specifications
- Antibody type:Primary
- Antigen name:CD4 antigen
- Antigen symbol:CD4
- Clonality:Monoclonal
- Clone:OKT-4
- Conjugation:iFluor® 594
- Flow cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:lgG2b kappa
- Reactivity:Human
- Cross adsorption:No
- Storage temperature:2...8 °C with minimised light exposure. Do not freeze.
- Shipping temperature:20
- Purification:Affinity chromatography
- Pack type:Vial
- Pk:100 Tests
Specifications
About this item
OKT-4 is an anti-human monoclonal antibody that targets the CD4 antigen.
- Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Caution: For research use only.
Type: Primary
Antigen: CD4 antigen
Clonality: Monoclonal
Clone: OKT-4
Conjugation: iFluor® 594
Epitope:
Host: Mouse
Isotype: Mouse igg2b, κ
Reactivity: Human