Anti-CD5 Mouse Monoclonal Antibody [clone: UCHT2]
Catalog # AATB10052000
Supplier: AAT Bioquest
Specifications
- Antibody type:Primary
- Antigen name:CD5 antigen
- Antigen symbol:CD5
- Clonality:Monoclonal
- Clone:UCHT2
- Flow cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Reactivity:Human
- Cross adsorption:No
- Storage temperature:Refrigerated 2...8 °C, minimise light exposure
- Shipping temperature:20
- Purification:Affinity chromatography
- Pack type:Vial
- Pk:100 µG
Specifications
About this item
UCHT2 is an anti-human monoclonal antibody that is specific for the CD5 antigen.
- Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Caution: For research use only.
Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: UCHT2
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human