Order Entry
Export
ContactUsLinkComponent
Anti-CD2 Mouse Monoclonal Antibody [clone: RPA-2.10]
Anti-CD2 Mouse Monoclonal Antibody [clone: RPA-2.10]
Catalog # AATB10021000
Supplier:  AAT Bioquest
undefined
Anti-CD2 Mouse Monoclonal Antibody [clone: RPA-2.10]
Catalog # AATB10021000
Supplier:  AAT Bioquest

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CD2 antigen
  • Antigen symbol:
    CD2
  • Clonality:
    Monoclonal
  • Clone:
    RPA-2.10
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Human
  • Cross adsorption:
    No
  • Storage temperature:
    Refrigerated 2...8 °C, minimise light exposure
  • Shipping temperature:
    20
  • Purification:
    Affinity chromatography
  • Pack type:
    Vial
  • Pk:
    100 µG

Specifications

About this item

RPA-2.10 is an anti-human monoclonal antibody that is specific for the CD2 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Caution: For research use only.

Type: Primary
Antigen: CD2
Clonality: Monoclonal
Clone: RPA-2.10
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human