Anti-CD8a Mouse Monoclonal Antibody [clone: SK1] (PacOrange)
Supplier: AAT BIOQUEST
Certificates
About this item
SK1 is an anti-human monoclonal antibody that is specific for the CD8a antigen.
- Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Caution: For research use only.
Type: Primary
Antigen: CD8a
Clonality: Monoclonal
Clone: SK1
Conjugation: PacOrange
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Specifications
- Catalog No:
- AATB100811K0
- AATB100811K1
- Antigen symbol:
- CD8a
- CD8a
- Antigen name:
- CD8a antigen
- CD8a antigen
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Pacific Orange
- Pacific Orange
- Clone:
- SK1
- SK1
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- ImmunoFluorescence:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Purification:
- Affinity chromatography
- Affinity chromatography
- Cross adsorption:
- No
- No
- Storage temperature:
- Refrigerated 2...8 °C, minimise light exposure
- Refrigerated 2...8 °C, minimise light exposure
- Shipping temperature:
- 20
- 20