Order Entry
Export
ContactUsLinkComponent
 
Anti-CD2 Mouse Monoclonal Antibody [clone: RPA-2.10] (PacOrange)
 
undefined
Anti-CD2 Mouse Monoclonal Antibody [clone: RPA-2.10] (PacOrange)

 

  • Catalog No:
  • AATB100211K0
  • AATB100211K1
  • Antigen symbol:
  • CD2
  • CD2
  • Antigen name:
  • CD2 antigen
  • CD2 antigen
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Pacific Orange
  • Pacific Orange
  • Clone:
  • RPA-2.10
  • RPA-2.10
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Purification:
  • Affinity chromatography
  • Affinity chromatography
  • Cross adsorption:
  • No
  • No
  • Storage temperature:
  • 2...8 °C with minimised light exposure, do not freeze
  • 2...8 °C with minimised light exposure, do not freeze
  • Shipping temperature:
  • 20
  • 20