Anti-CD161 Mouse Monoclonal Antibody [clone: HP-3G10] (mFluor™ Violet 450)
Supplier: AAT Bioquest
Certificates
About this item
HP-3G10 is an anti-human monoclonal antibody that recognises the CD161 antigen.
- Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Caution: For research use only.
Type: Primary
Antigen: CD161
Clonality: Monoclonal
Clone: HP-3G10
Conjugation: mFluor™ Violet 450
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Specifications
- Catalog No:
- AATB116100Z1
- AATB116100Z0
- Antigen symbol:
- CD161
- CD161
- Antigen name:
- CD161 antigen
- CD161 antigen
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- mFluor™ Violet 450
- mFluor™ Violet 450
- Clone:
- HP-3G10
- HP-3G10
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- ImmunoFluorescence:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Purification:
- Affinity chromatography
- Affinity chromatography
- Cross adsorption:
- No
- No
- Storage temperature:
- 2...8 °C with minimised light exposure, do not freeze
- 2...8 °C with minimised light exposure, do not freeze
- Shipping temperature:
- 20
- 20