Anti-CD27 Mouse Monoclonal Antibody [clone: O323] (iFluor® 647)
Catalog # AATB102710F0
Supplier: AAT Bioquest
Specifications
- Antibody type:Primary
- Antigen name:CD27 antigen
- Antigen symbol:CD27
- Clonality:Monoclonal
- Clone:O323
- Conjugation:iFluor® 647
- Flow cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:lgG1 kappa
- Reactivity:Human
- Cross adsorption:No
- Storage temperature:2...8 °C with minimised light exposure, do not freeze
- Shipping temperature:20
- Purification:Affinity chromatography
- Pack type:Vial
- Pk:100 Tests
Specifications
About this item
O323 is an anti-human monoclonal antibody that is specific for the CD27 antigen.
- Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Caution: For research use only.
Type: Primary
Antigen: CD27
Clonality: Monoclonal
Clone: O323
Conjugation: iFluor® 647
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human