Order Entry
Export
ContactUsLinkComponent
Anti-CD5 Mouse Monoclonal Antibody [clone: UCHT2] (iFluor® 633)
Anti-CD5 Mouse Monoclonal Antibody [clone: UCHT2] (iFluor® 633)
Supplier:  AAT BIOQUEST
Certificates
undefined
Anti-CD5 Mouse Monoclonal Antibody [clone: UCHT2] (iFluor® 633)
Supplier:  AAT BIOQUEST

Specifications

  • Catalog No:
  • AATB100520E0
  • AATB100520E1
  • Antigen symbol:
  • CD5
  • CD5
  • Antigen name:
  • CD5 antigen
  • CD5 antigen
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • iFluor® 633
  • iFluor® 633
  • Clone:
  • UCHT2
  • UCHT2
  • Reactivity:
  • Human
  • Host:
  • Mouse
  • Mouse
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Purification:
  • Affinity chromatography
  • Affinity chromatography
  • Cross adsorption:
  • No
  • No
  • Storage temperature:
  • 2...8 °C with minimised light exposure, do not freeze
  • 2...8 °C with minimised light exposure, do not freeze
  • Shipping temperature:
  • 20
  • 20

Specifications