Anti-SLC25A52 Rabbit Polyclonal Antibody
Catalog # PRSI55-491
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Solute carrier family 25 member 52
- Antigen symbol:SLC25A52
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- Western blot:Yes
- Epitope:Amino Acid Range = 46-75
- Form:Liquid
- Gene ID:Q3SY17
- Antigen synonyms:SLC25A52
- Modification:None
- Storage buffer:Supplied in PBS with 0,09% (W/V) sodium azide
- Molecular weight:34 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This MCART2 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 46-75 amino acids from the N-terminal region of human MCART2.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Size:400 µl
- Pk:400 µl
Specifications
About this item
This gene is similar to the mitochondrial carrier triple repeat 1 gene on chromosome 9. The gene is intronless and may be an evolving pseudogene; however, it is transcribed and it contains a full-length coding region so it is currently classified as a protein-coding locus.
For WB starting dilution is: 1:1000. For FACS starting dilution is: 1:10~50.
Type: Primary
Antigen: SLC25A52
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human