Anti-PRDM10 Rabbit Polyclonal Antibody
Catalog # PRSI15-914
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:PR domain containing 10
- Antigen symbol:PRDM10
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Form:Liquid
- Gene ID:56980
- Antigen synonyms:KIAA1231|PFM7|PR domain containing 10|tristanin|TRIS|PRDM10
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 600-870 of human PRDM10 (NP_064613.2)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: PRDM10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human