Anti-KARS Rabbit Polyclonal Antibody
Vial
Catalog # PRSI15-194
Supplier: ProSci Inc.
Jump to:
Product Details & Documents
Type: Primary
Antigen: KARS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Pack type:Vial
- Western blot:Yes
- Clonality:Polyclonal
- Modification:None
- Purification:Affinity purification
- Antibody type:Primary
- Form:Liquid
- Isotype:IgG
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles
- Host:Rabbit
- Concentration:Batch dependent
- Shipping temperature:20
- Antigen name:Lysyl-tRNA synthetase
- Cat. No.:PRSI15-194
- Reactivity:Human|Rat|Mouse
- Gene ID:3735
- Antigen synonyms:KRS|KARS2|KARS|CMTRIB|DFNB89|KARS1
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 298-597 of human KARS (NP_005539.1)
- Conjugation:Unconjugated
- Antigen symbol:KARS
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3



