Order Entry
Export
Orders LinkContactUsLinkComponent
Anti-TWF2 Rabbit Polyclonal Antibody

Anti-TWF2 Rabbit Polyclonal Antibody

Vial
Catalog # PRSI15-123
Supplier:   ProSci Inc.
Anti-TWF2 Rabbit Polyclonal Antibody
Catalog # PRSI15-123
Supplier:  ProSci Inc.
Anti-TWF2 Rabbit Polyclonal Antibody

Vial

Price
484,00 €
Log in for availability
1 * 50 µl
Product Details & Documents

Type: Primary
Antigen: TWF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse

Specifications
  • Pack type:
    Vial
  • Western blot:
    Yes
  • Clonality:
    Polyclonal
  • Modification:
    None
  • ImmunoPrecipitation:
    Yes
  • Purification:
    Affinity purification
  • Antibody type:
    Primary
  • Form:
    Liquid
  • Isotype:
    IgG
  • Storage temperature:
    Store at −20 °C. Avoid freeze/thaw cycles
  • Host:
    Rabbit
  • Concentration:
    Batch dependent
  • Shipping temperature:
    20
  • Antigen name:
    Twinfilin-2
  • Cat. No.:
    PRSI15-123
  • Reactivity:
    Human|Mouse
  • Gene ID:
    11344
  • Antigen synonyms:
    A6-related protein|TWF2|Twinfilin-1-like protein|Twinfilin-2|Protein tyrosine kinase 9-like|PTK9L|hA6RP
  • Immunogen:
    Recombinant fusion protein containing a sequence corresponding to amino acids 1-369 of human TWF2 (NP_009215.1)
  • ImmunoChemistry:
    Yes
  • Conjugation:
    Unconjugated
  • Antigen symbol:
    TWF2
  • Storage buffer:
    PBS with 0,02% sodium azide, 50% glycerol, pH 7,3